accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CSSNNH3C

CAS RN 130-61-0

IUPAC Name: hydrogen 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-(methylsulfanyl)-10H-phenothiazine chloride
Synonyms: thioridazine hydrochloride thioridazine chloride thioridazine hcl ridazine mellaril hydrochloride sonapax hydrochloride ridazin 利达嗪 aldazine melleretten
Molecular Weight (g/mol): 407.03
Molecular Formula: C21H27ClN2S2
InChi Key: NZFNXWQNBYZDAQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CSC1=CC=C2SC3=CC=CC=C3N(CCC3CCCCN3C)C2=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results