accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClH3CH3CNNCH3CH3

CAS RN 130065-61-1

IUPAC Name: dihydrogen (3-{8,8-dipropyl-2-azaspiro[4.5]decan-2-yl}propyl)diethylamine dichloride
Synonyms: atiprimod
Molecular Weight (g/mol): 409.52
Molecular Formula: C22H46Cl2N2
InChi Key: MOUZYBYTICOTFQ-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCCC1(CCC)CCC2(CCN(CCCN(CC)CC)C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results