CAS RN 130100-20-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Mag-Fura-2 AM
Magnesium (Mg2+) indicator, membrane permeable