accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHO(S)NHO(R)CH3CH3(S)SSOHNOH

CAS RN 130370-60-4

IUPAC Name: (2R,3S)-N'-hydroxy-N-[(1S)-1-(methylcarbamoyl)-2-phenylethyl]-2-(2-methylpropyl)-3-[(thiophen-2-ylsulfanyl)methyl]butanediamide
Synonyms: batimastat 巴马司他
Molecular Weight (g/mol): 477.64
Molecular Formula: C23H31N3O4S2
InChi Key: XFILPEOLDIKJHX-QYZOEREBSA-N
SMILES: CNC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@H](CC(C)C)[C@H](CSC1=CC=CS1)C(=O)NO
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results