CAS RN 130465-39-3
IUPAC Name:
1-(aminomethyl)-3-phenyl-3,4-dihydro-1H-2-benzopyran-5,6-diol hydrochloride
Synonyms:
1-(aminomethyl)-3-phenyl-3,4-dihydro-1H-2-benzopyran-5,6-diol hydrochloride
Molecular Weight (g/mol):
307.77
Molecular Formula:
C16H18ClNO3
InChi Key:
PQPGUUQPTSMLKU-UHFFFAOYNA-N
SMILES:
Cl.NCC1OC(CC2=C1C=CC(O)=C2O)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ A 68930 hydrochloride
CAS: 130465-39-3 Molecular Formula: C16H18ClNO3