accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NONHOHO

CAS RN 13059-60-4

IUPAC Name: hydrogen 2-(2-aminoacetamido)acetic acid chloride
Synonyms: dipeptide-15
Molecular Weight (g/mol): 168.58
Molecular Formula: C4H9ClN2O3
InChi Key: YHBAZQDEMYQPJL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results