accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOClSOOO

CAS RN 13064-50-1

IUPAC Name: sodium 2-chloro-1-hydroxyethane-1-sulfonate
Synonyms: sodium 2-chloro-1-hydroxyethanesulfonate
Molecular Weight (g/mol): 182.55
Molecular Formula: C2H4ClNaO4S
InChi Key: FOSIEWVVVRVFJH-UHFFFAOYNA-M
SMILES: [Na+].OC(CCl)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results