Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetaldehyde Sodium Bisulfite 95.0+%, TCI America™
SDP

Catalog No. C074925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C074925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C074925G Supplier TCI America Supplier No. C074925G

Chemical Identifiers

CAS 13064-50-1
Molecular Formula C2H4ClNaO4S
Molecular Weight (g/mol) 182.55
MDL Number MFCD00059901
InChI Key FOSIEWVVVRVFJH-UHFFFAOYNA-M
Synonym 2-Chloroethanol-1-sulfonic Acid Sodium Salt, 2-Chloro-1-hydroxyethanesulfonic Acid Sodium Salt, Sodium 2-Chloro-1-hydroxyethanesulfonate
PubChem CID 88666897
IUPAC Name sodium 2-chloro-1-hydroxyethane-1-sulfonate
SMILES [Na+].OC(CCl)S([O-])(=O)=O

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 182.55
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloroacetaldehyde Sodium Bisulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.