CAS RN 130663-39-7
IUPAC Name:
1-{[4-(dimethylamino)-3-methylphenyl]methyl}-5-(2,2-diphenylacetyl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine-6-carboxylic acid
Synonyms:
1-{[4-(dimethylamino)-3-methylphenyl]methyl}-5-(2,2-diphenylacetyl)-4H,6H,7H-imidazo[4,5-c]pyridine-6-carboxylic acid
Molecular Weight (g/mol):
508.62
Molecular Formula:
C31H32N4O3
InChi Key:
YSTVFDAKLDMYCR-UHFFFAOYNA-N
SMILES:
CN(C)C1=CC=C(CN2C=NC3=C2CC(N(C3)C(=O)C(C2=CC=CC=C2)C2=CC=CC=C2)C(O)=O)C=C1C
1
–
1
of
1
results
1
R&D Systems™ PD 123319 ditrifluoroacetate
CAS: 130663-39-7 Molecular Formula: C33H33F3N4O5