CAS RN 1306761-53-4
IUPAC Name:
3-[(4-{5-[3-cyano-4-(propan-2-yloxy)phenyl]-1,2,4-oxadiazol-3-yl}-2,3-dihydro-1H-inden-1-yl)amino]propanoic acid hydrochloride
Synonyms:
3-({4-[5-(3-cyano-4-isopropoxyphenyl)-1,2,4-oxadiazol-3-yl]-2,3-dihydro-1H-inden-1-yl}amino)propanoic acid hydrochloride
Molecular Weight (g/mol):
468.94
Molecular Formula:
C24H25ClN4O4
InChi Key:
ZHPGSGYEFGEZAL-UHFFFAOYNA-N
SMILES:
Cl.CC(C)OC1=C(C=C(C=C1)C1=NC(=NO1)C1=C2CCC(NCCC(O)=O)C2=CC=C1)C#N
1
–
1
of
1
results
1
R&D Systems™ RP 001 hydrochloride
CAS: 1306761-53-4 Molecular Formula: C24H25ClN4O4