CAS RN 130722-95-1
IUPAC Name:
3-(benzyloxy)-5-bromopyridine
Synonyms:
3-(benzyloxy)-5-bromopyridine
Molecular Weight (g/mol):
264.12
Molecular Formula:
C12H10BrNO
InChi Key:
YSHKYZAWTWKQKK-UHFFFAOYSA-N
SMILES:
BrC1=CN=CC(OCC2=CC=CC=C2)=C1
1
–
1
of
1
results
1
3-Benzyloxy-5-bromopyridine 98.0+%, TCI America™
CAS: 130722-95-1 Molecular Formula: C12H10BrNO Molecular Weight (g/mol): 264.122 MDL Number: MFCD07375004 InChI Key: YSHKYZAWTWKQKK-UHFFFAOYSA-N PubChem CID: 15389198 IUPAC Name: 3-bromo-5-phenylmethoxypyridine SMILES: C1=CC=C(C=C1)COC2=CC(=CN=C2)Br