accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130800-90-7

IUPAC Name: 2-(4-methylpiperazin-1-yl)-5-(2,3,5-trichlorophenyl)pyrimidin-4-amine
Synonyms: 2-(4-methylpiperazin-1-yl)-5-(2,3,5-trichlorophenyl)pyrimidin-4-amine
Molecular Weight (g/mol): 372.68
Molecular Formula: C15H16Cl3N5
InChi Key: PDOCBJADCWMDGL-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=NC=C(C(N)=N1)C1=CC(Cl)=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results