accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOHOOCH3

CAS RN 13081-97-5

IUPAC Name: 3-hydroxy-2-(hydroxymethyl)-2-[(octadecanoyloxy)methyl]propyl octadecanoate
Synonyms: pentaerythrityl distearate
Molecular Weight (g/mol): 669.09
Molecular Formula: C41H80O6
InChi Key: FSEJJKIPRNUIFL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(CO)(CO)COC(=O)CCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results