accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130812-47-4

IUPAC Name: barium(2+) bis(2-ethoxyethan-1-olate)
Synonyms: barium(2+) bis(2-ethoxyethanolate)
Molecular Weight (g/mol): 315.56
Molecular Formula: C8H18BaO4
InChi Key: WCXRGQPXPCQKSJ-UHFFFAOYSA-N
SMILES: [Ba++].CCOCC[O-].CCOCC[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results