Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium 1-methoxy-2-propoxide, 25% w/w in 1-methoxy-2-propanol, Thermo Scientific™

Catalog No. AA4125714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4125714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4125714 Supplier Thermo Scientific Chemicals Supplier No. 4125714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130812-47-4
Molecular Formula C8H18BaO4
Molecular Weight (g/mol) 315.555
MDL Number MFCD00078021
InChI Key WCXRGQPXPCQKSJ-UHFFFAOYSA-N
Synonym bis 2-ethoxyethoxy barium, barium methoxypropoxide w/w in methoxypropanol, barium bis 2-ethoxyethan-1-olate, barium 2+ ion bis 2-ethoxyethanolate
PubChem CID 14619883
IUPAC Name barium(2+);2-ethoxyethanolate
SMILES CCOCC[O-].CCOCC[O-].[Ba+2]

Specifications

Density 1.04 g/mL
Flash Point 35°C (95°F)
Linear Formula Ba[OCH(CH3)CH2OCH3]2
Quantity 25 g
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 315.55
Concentration or Composition (by Analyte or Components) 25% w/w in 1-methoxy-2-propanol
Physical Form Liquid
Chemical Name or Material Barium 1-methoxy-2-propoxide
Hazard Category H226-H314-H335-H336
Hazard Statement GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.