CAS RN 1309976-62-2
IUPAC Name:
N-cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]pyridine-3-carboxamide hydrochloride
Synonyms:
N-cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]pyridine-3-carboxamide hydrochloride
Molecular Weight (g/mol):
330.79
Molecular Formula:
C18H16ClFN2O
InChi Key:
NBGAPTWZQXSEAA-UHFFFAOYSA-N
SMILES:
Cl.FC1=CC=CC(=C1)C#CC1=NC=C(C=C1)C(=O)NC1CCC1
1
–
1
of
1
results
1
R&D Systems™ VU 0360172 hydrochloride
CAS: 1309976-62-2 Molecular Formula: C18H16ClFN2O