accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131-69-1

IUPAC Name: 2-{[4-(acetamidosulfonyl)phenyl]carbamoyl}benzoic acid
Synonyms: tamid phthalylsulfacetamide
Molecular Weight (g/mol): 362.36
Molecular Formula: C16H14N2O6S
InChi Key: SNWQKAWITMVCQW-UHFFFAOYSA-N
SMILES: CC(=O)NS(=O)(=O)C1=CC=C(NC(=O)C2=CC=CC=C2C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)