accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13101-54-7

IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: erlose sucrosylglucose glucosylsucrose
Molecular Weight (g/mol): 504.44
Molecular Formula: C18H32O16
InChi Key: FVVCFHXLWDDRHG-KKNDGLDKSA-N
SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results