CAS RN 131123-76-7
IUPAC Name:
5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
Synonyms:
5,7-dichlorokynurenic acid
Molecular Weight (g/mol):
258.05
Molecular Formula:
C10H5Cl2NO3
InChi Key:
BGKFPRIGXAVYNX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=O)C2=C(Cl)C=C(Cl)C=C2N1
1
–
1
of
1
results
1
R&D Systems™ 5,7-Dichlorokynurenic acid
CAS: 131123-76-7 Molecular Formula: C10H5Cl2NO3