accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1312473-63-4

IUPAC Name: 4-[4-(2-fluoro-4-methoxyphenyl)-2-oxo-1,2-dihydropyridin-1-yl]-N-hydroxy-2-methanesulfonyl-2-methylbutanamide
Synonyms: 4-[4-(2-fluoro-4-methoxyphenyl)-2-oxopyridin-1-yl]-N-hydroxy-2-methanesulfonyl-2-methylbutanamide
Molecular Weight (g/mol): 412.43
Molecular Formula: C18H21FN2O6S
InChi Key: DNVUWHWBCMGQLU-UHFFFAOYNA-N
SMILES: COC1=CC(F)=C(C=C1)C1=CC(=O)N(CCC(C)(C(=O)NO)S(C)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results