Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF 5081090, Tocris Bioscience™
SDP

Catalog No. 436210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-6210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-6210 Supplier R&D Systems Supplier No. 4362/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent LpxC inhibitor

Potent antibacterial agent which disrupts lipid bilayer synthesis through inhibition of LpxC (IC 50 = 1.1 nM in Pseudomonas aeruginosa); effective against a range of gram negative bacteria.

Chemical Identifiers

CAS 1312473-63-4
Molecular Formula C18H21FN2O6S
Molecular Weight (g/mol) 412.432
InChI Key DNVUWHWBCMGQLU-GOSISDBHSA-N
Synonym 2r-4-4-2-fluoro-4-methoxyphenyl-2-oxopyridin-1 2h-yl-n-hydroxy-2-methyl-2-methylsulfonyl butanamide, 8gj, pf hplc, ?r-4-2-fluoro-4-methoxyphenyl-n-hydroxy-?-methyl-?-methylsulfonyl-2-oxo-1 2h-pyridinebutanamide, ar-4-2-fluoro-4-methoxyphenyl-n-hydroxy-, a-methyl-, a-methylsulfonyl-2-oxo-1 2h-pyridinebutanamide, 2r-4-4-2-fluoro-4-methoxyphenyl-2-oxopyridin-1 2h-yl-n-hydroxy-2-methanesulfonyl-2-methylbutanamide, 2r-4-4-2-fluoro-4-methoxyphenyl-2-oxopyridin-1-yl-n-hydroxy-2-methanesulfonyl-2-methylbutanamide
PubChem CID 56964907
IUPAC Name (2R)-4-[4-(2-fluoro-4-methoxyphenyl)-2-oxopyridin-1-yl]-N-hydroxy-2-methyl-2-methylsulfonylbutanamide
SMILES CC(CCN1C=CC(=CC1=O)C2=C(C=C(C=C2)OC)F)(C(=O)NO)S(=O)(=O)C

Specifications

Quantity 10 mg
Formula Weight 412.43
Percent Purity >98%
Chemical Name or Material PF 5081090
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.