accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1312992-24-7

IUPAC Name: disodium 6,7-dinitroquinoxaline-2,3-bis(olate)
Synonyms: disodium 6,7-dinitroquinoxaline-2,3-bis(olate)
Molecular Weight (g/mol): 296.11
Molecular Formula: C8H2N4Na2O6
InChi Key: GPSBSOYURFUVKJ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=NC2=CC(=C(C=C2N=C1[O-])[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results