accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHNSSONHONOHNOHNNH2NNH2NNH2H2NH2NOHNH2NOHNNHNONHSSHNONHONHONOHNONH2NNH2NH2NNH2H3CCH3OOH

CAS RN 1313027-43-8

IUPAC Name: 14-[2-(6-amino-2-{[14-amino-3,17-bis({3-[(diaminomethylidene)amino]propyl})-6-(2-methylpropyl)-1,4,7,15,18-pentaoxo-icosahydropyrrolo[2,1-m]1,2-dithia-5,8,11,14,17-pentaazacycloicosan-9-yl]formamido}hexanamido)-3-(1H-imidazol-5-yl)propanamido]-3,17-bis({3-[(diaminomethylidene)amino]propyl})-6-(2-methylpropyl)-1,4,7,15,18-pentaoxo-icosahydropyrrolo[2,1-m]1,2-dithia-5,8,11,14,17-pentaazacycloicosane-9-carboxylic acid
Synonyms: 14-[2-(6-amino-2-{[14-amino-3,17-bis({3-[(diaminomethylidene)amino]propyl})-6-(2-methylpropyl)-1,4,7,15,18-pentaoxo-tetradecahydro-2H-pyrrolo[2,1-m]1,2-dithia-5,8,11,14,17-pentaazacycloicosan-9-yl]formamido}hexanamido)-3-(3H-imidazol-4-yl)propanamido]-3,17-bis({3-[(diaminomethylidene)amino]propyl})-6-(2-methylpropyl)-1,4,7,15,18-pentaoxo-tetradecahydro-2H-pyrrolo[2,1-m]1,2-dithia-5,8,11,14,17-pentaazacycloicosane-9-carboxylic acid
Molecular Weight (g/mol): 1737.17
Molecular Formula: C70H121N29O15S4
InChi Key: BMJYNYGARUVBJU-UHFFFAOYNA-N
SMILES: CC(C)CC1NC(=O)C(CCCN=C(N)N)NC(=O)C2CCCN2C(=O)C(CCCN=C(N)N)NC(=O)C(CSSCC(NC1=O)C(O)=O)NC(=O)C(CC1=CN=CN1)NC(=O)C(CCCCN)NC(=O)C1CSSCC(N)C(=O)NC(CCCN=C(N)N)C(=O)N2CCCC2C(=O)NC(CCCN=C(N)N)C(=O)NC(CC(C)C)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)