Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MM 54
GREENER_CHOICESDP

Catalog No. 59921 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-921 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-921 Supplier Tocris Bioscience™ Supplier No. 5992/1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent apelin receptor antagonist

Potent apelin receptor antagonist (Ki = 82 nM; IC50 = 93 nM). Antagonizes the inhibitory affect of [Pyr1]-Apelin-13 on forskolin-induced cAMP accumulation in CHO-K1-APJ cells. Recuces tumor expansion and lengthens survival time in a mouse xenograft model of glioblastoma.

Chemical Identifiers

CAS 1313027-43-8

Specifications

Purity 95%
Content And Storage Store at -20°C
Molecular Weight (g/mol) 1737.166
Quantity 1 mg
Solubility Soluble to 2mg/mL in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.