accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNHNHNONO

CAS RN 1316652-41-1

IUPAC Name: N-oxo-3-{1-[(phenylsulfanyl)methyl]-2,3-dihydro-1H-1,2,3-triazol-4-yl}benzamide
Synonyms: N-oxo-3-{1-[(phenylsulfanyl)methyl]-2,3-dihydro-1,2,3-triazol-4-yl}benzamide
Molecular Weight (g/mol): 326.37
Molecular Formula: C16H14N4O2S
InChi Key: YMCCQRYCIRCIGS-UHFFFAOYSA-N
SMILES: O=NC(=O)C1=CC=CC(=C1)C1=CN(CSC2=CC=CC=C2)NN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results