CAS RN 1317-36-8
Lead(II) oxide, 99.9%
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: oxolead SMILES: O=[Pb]
Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) oxide, 99+%
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: plumbanone SMILES: O=[Pb]
Lead(II) oxide, Puratronic™, 99.9995% (metals basis)
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) Oxide, Extra Pure, MilliporeSigma™
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: plumbanone SMILES: O=[Pb]
Lead(II) Oxide, For Analysis, EMSURE™, MilliporeSigma™
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: plumbanone SMILES: O=[Pb]
Lead(II) oxide, low silver
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(II) oxide, Puratronic™, 99.999% (metals basis)
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: oxolead SMILES: O=[Pb]
Lead(II) oxide, 99.99% (metals basis)
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Lead(Ii) Oxide, puriss. p.a., ≥99.0% (KT), Solstice
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 SMILES: O=[Pb]
Sigma Aldrich Lead(II) oxide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Lead Monoxide, MP Biomedicals
CAS: 1317-36-8 Molecular Formula: OPb Molecular Weight (g/mol): 223.20 MDL Number: MFCD00011164 InChI Key: YEXPOXQUZXUXJW-UHFFFAOYSA-N Synonym: lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow PubChem CID: 14827 IUPAC Name: plumbanone SMILES: O=[Pb]