Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Oxide, Yellow Powder, 95%, Spectrum™ Chemical
SDP

Catalog No. 18611251 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-251 500 g Fiber Drum
18-611-249 125 g Fiber Drum
18-611-248 2.5 kg Fiber Drum
18-611-250 12 kg Fiber Drum
18-611-247 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-251 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1080500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1317-36-8

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 500 g
Molecular Weight (g/mol) 223.19
Percent Purity 95%
Grade Reagent
Packaging Fiber Drum
Chemical Name or Material Lead Oxide, Yellow, Powder,

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.