Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(Ii) Oxide, puriss. p.a., ≥99.0% (KT), Solstice

Catalog No. 60044873 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-873 1 kg Poly Bottle
60-044-874 250 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-873 Supplier Solstice Supplier No. 153381KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.0% (KT), yellow

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Boiling Point 1470°C
Melting Point 890°C
Color Yellow
Physical Form Solid
Quantity 1 kg
Packaging Poly Bottle
Vapor Pressure 10mmHg (0°C)
Density 9.66g/cu.cm
Linear Formula PbO
UN Number UN2291
IUPAC Name plumbanone
Formula Weight 223.2g/mol
Percent Purity ≥99.0% (KT)
Grade puriss. p.a.
Chemical Name or Material Lead(Ii) Oxide
Show More Show Less
Hazard Statement P201-P260-P263-P280-P301 + P312 + P330-P308 + P313
Precautionary Statement H302 + H332-H351-H360Df-H362-H372-H410
DOTInformation 6.1/PGIII
EINECSNumber 215-267-0
RTECSNumber OG1750000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.