accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrHO

CAS RN 131747-54-1

IUPAC Name: (2-bromopyridin-3-yl)methanol
Synonyms: (2-bromopyridin-3-yl)methanol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: QIQZUABMLGJKJP-UHFFFAOYSA-N
SMILES: OCC1=C(Br)N=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results