CAS RN 132-65-0
Dibenzothiophene, 98%
CAS: 132-65-0 Molecular Formula: C12H8S Molecular Weight (g/mol): 184.26 MDL Number: MFCD00004969 InChI Key: IYYZUPMFVPLQIF-UHFFFAOYSA-N Synonym: diphenylene sulfide,dibenzo b,d thiophene,9-thiafluorene,alpha-thiafluorene,2,2'-biphenylylene sulfide,usan,unii-z3d4aj1r48,1,1'-biphenyl-2,2'-diyl sulfide,.alpha.-thiafluorene PubChem CID: 3023 ChEBI: CHEBI:23681 IUPAC Name: dibenzothiophene SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3S2
Dibenzothiophene 98.0+%, TCI America™
CAS: 132-65-0 Molecular Formula: C12H8S Molecular Weight (g/mol): 184.256 MDL Number: MFCD00004969 InChI Key: IYYZUPMFVPLQIF-UHFFFAOYSA-N Synonym: diphenylene sulfide,dibenzo b,d thiophene,9-thiafluorene,alpha-thiafluorene,2,2'-biphenylylene sulfide,usan,unii-z3d4aj1r48,1,1'-biphenyl-2,2'-diyl sulfide,.alpha.-thiafluorene PubChem CID: 3023 ChEBI: CHEBI:23681 IUPAC Name: dibenzothiophene SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3S2
Dibenzothiophene, 98%
CAS: 132-65-0 Molecular Formula: C12H8S Molecular Weight (g/mol): 184.256 MDL Number: MFCD00004969 InChI Key: IYYZUPMFVPLQIF-UHFFFAOYSA-N Synonym: diphenylene sulfide,dibenzo b,d thiophene,9-thiafluorene,alpha-thiafluorene,2,2'-biphenylylene sulfide,usan,unii-z3d4aj1r48,1,1'-biphenyl-2,2'-diyl sulfide,.alpha.-thiafluorene PubChem CID: 3023 ChEBI: CHEBI:23681 IUPAC Name: dibenzothiophene SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3S2
Sigma Aldrich Dibenzothiophene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dibenzothiophene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Dibenzothiophene, SPEX CertiPrep™
CAS: 132-65-0 Molecular Formula: C12H8S Molecular Weight (g/mol): 184.256 InChI Key: IYYZUPMFVPLQIF-UHFFFAOYSA-N PubChem CID: 3023 ChEBI: CHEBI:23681 IUPAC Name: dibenzothiophene SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3S2