Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene, SPEX CertiPrep™
SDP

Catalog No. S1185 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S-1185 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S-1185 Supplier SPEX CertiPrep Supplier No. S1185

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Dibenzothiophene132-65-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 132-65-0 , 67-56-1
Molecular Formula C12H8S
Molecular Weight (g/mol) 184.256
InChI Key IYYZUPMFVPLQIF-UHFFFAOYSA-N
PubChem CID 3023
ChEBI CHEBI:23681
IUPAC Name dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Chemical Name or Material Dibenzothiophene
Hazard Category Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Fatal in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause genetic defects. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.