accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1320-06-5

IUPAC Name: (1Z)-1-(2-{4-[2-(2,5-dimethylphenyl)diazen-1-yl]-2,5-dimethylphenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Synonyms: oil red O
Molecular Weight (g/mol): 408.51
Molecular Formula: C26H24N4O
InChi Key: HJAYEODIXUYVIC-LTFAAIOASA-N
SMILES: CC1=CC=C(C)C(=C1)N=NC1=CC(C)=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1C
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Color 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)