accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13200-85-6

OHOHNO

CAS RN 13200-85-6

IUPAC Name: 2-formamido-3-phenylpropanoic acid
Synonyms: 2-formamido-3-phenylpropanoic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: NSTPXGARCQOSAU-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)