accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNN

CAS RN 13228-40-5

IUPAC Name: 2-(pyridin-2-yl)-1H-indole
Synonyms: 2-(2-pyridyl)indole
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: OLGGLCIDAMICTA-UHFFFAOYSA-N
SMILES: N1C(=CC2=CC=CC=C12)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results