accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOCH3HOHO

CAS RN 1323-39-3

IUPAC Name: octadecanoic acid; propane-1,2-diol
Synonyms: 1,2-propanediol; stearic acid
Molecular Weight (g/mol): 360.58
Molecular Formula: C21H44O4
InChi Key: CMCJFUXWBBHIIL-UHFFFAOYNA-N
SMILES: CC(O)CO.CCCCCCCCCCCCCCCCCC(O)=O
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)