Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol Monostearate (contains ca. 35% Monopalmitate), TCI America™
SDP

Catalog No. P061425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P061425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P061425G Supplier TCI America Supplier No. P061425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1323-39-3
Molecular Formula C21H42O3
MDL Number MFCD00021902
Synonym Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester

Specifications

Color White
Quantity 25 g
Formula Weight 342.56
Physical Form Crystalline Powder
Chemical Name or Material Propylene Glycol Monostearate (contains ca. 35% Monopalmitate)
EINECSNumber (2)-0772&(2)-2523
RTECSNumber WI4550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.