accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONN

CAS RN 132521-82-5

IUPAC Name: methyl 6-(piperidin-1-yl)pyridine-3-carboxylate
Synonyms: methyl 6-(piperidin-1-yl)pyridine-3-carboxylate
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: UJSPHYCMAZIKQD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results