accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3CH3HO

CAS RN 13254-34-7

IUPAC Name: 2,6-dimethylheptan-2-ol
Synonyms: 2-heptanol, 2,6-dimethyl-
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: HGDVHRITTGWMJK-UHFFFAOYSA-N
SMILES: CC(C)CCCC(C)(C)O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)