Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-2-heptanol, 99%, Thermo Scientific™

Catalog No. AAB2569118
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2569118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2569118 Supplier Thermo Scientific Chemicals Supplier No. B2569118

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13254-34-7
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00072198
InChI Key HGDVHRITTGWMJK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-2-heptanol, 2-heptanol, 2,6-dimethyl, dimetol, freesiol, lolitol, dimethyl heptanol, acmc-20ao86, dimetol dimethyl heptanol, dsstox_cid_21424, dsstox_rid_79728
PubChem CID 83268
IUPAC Name 2,6-dimethylheptan-2-ol
SMILES CC(C)CCCC(C)(C)O

Specifications

Melting Point -10°C
Density 0.81
Boiling Point 179°C to 181°C
Flash Point 63°C (145°F)
Odor Characteristic
Refractive Index 1.426
Quantity 50 g
Formula Weight 144.25
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-2-heptanol
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 236-244-1
RTECSNumber MJ3324950
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.