Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-2-heptanol, 99%, Thermo Scientific™

Catalog No. AC259645000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC259645000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259645000 Supplier Thermo Scientific Chemicals Supplier No. 259645000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13254-34-7
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
InChI Key HGDVHRITTGWMJK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-2-heptanol, 2-heptanol, 2,6-dimethyl, dimetol, freesiol, lolitol, dimethyl heptanol, acmc-20ao86, dimetol dimethyl heptanol, dsstox_cid_21424, dsstox_rid_79728
PubChem CID 83268
IUPAC Name 2,6-dimethylheptan-2-ol
SMILES CC(C)CCCC(C)(C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.81g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Quantity 500 mL
Formula Weight 144.26
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethyl-2-heptanol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.