accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13280-61-0

IUPAC Name: 1-methyl-2-[(1E)-2-{4-[(1E)-2-(2-methylphenyl)ethenyl]phenyl}ethenyl]benzene
Synonyms: 1-methyl-2-[(1E)-2-{4-[(1E)-2-(2-methylphenyl)ethenyl]phenyl}ethenyl]benzene
Molecular Weight (g/mol): 310.44
Molecular Formula: C24H22
InChi Key: QKLPIYTUUFFRLV-YTEMWHBBSA-N
SMILES: CC1=CC=CC=C1\C=C\C1=CC=C(\C=C\C2=CC=CC=C2C)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)