Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(2-methylstyryl)benzene, 99%

Catalog No. AAA1353406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1353406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1353406 Supplier Thermo Scientific Chemicals Supplier No. A1353406
  • $90.00
    $76.65 / Each of 1

    Save $13.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 13280-61-0 | C24H22 | 310.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13280-61-0
Molecular Formula C24H22
Molecular Weight (g/mol) 310.44
MDL Number MFCD00008529
InChI Key QKLPIYTUUFFRLV-YTEMWHBBSA-N
Synonym 1,4-bis 2-methylstyryl benzene, bis-msb, p-bis o-methylstyryl benzene, benzene, 1,4-bis 2-2-methylphenyl ethenyl, 1,4-di 2-methylstyryl benzene, 1,4-bis 2-methylstyryl benzene bis-msb, benzene, p-bis o-methylstyryl, 1,4-di-2-methylstyryl benzene, 2-menthylstyrene, 1-methyl-2-2-4-2-2-methylphenyl ethenyl phenyl ethenyl benzene
PubChem CID 5378735
IUPAC Name 1,4-bis[(E)-2-(2-methylphenyl)ethenyl]benzene
SMILES CC1=CC=CC=C1C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3C

Specifications

Melting Point 179°C to 184°C
Odor Odorless
Quantity 5 g
Beilstein 2375845
Sensitivity Light sensitive
Formula Weight 310.44
Percent Purity 99%
Chemical Name or Material 1,4-Bis(2-methylstyryl)benzene
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
EINECSNumber 236-285-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.