Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(2-methylstyryl)benzene 99.0+%, TCI America™
SDP

Catalog No. B10245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10245G 5 g
B102425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10245G Supplier TCI America Supplier No. B10245G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13280-61-0
Molecular Formula C24H22
Molecular Weight (g/mol) 310.44
MDL Number MFCD00008529
InChI Key QKLPIYTUUFFRLV-YTEMWHBBSA-N
Synonym 1,4-bis 2-methylstyryl benzene, bis-msb, p-bis o-methylstyryl benzene, benzene, 1,4-bis 2-2-methylphenyl ethenyl, 1,4-di 2-methylstyryl benzene, 1,4-bis 2-methylstyryl benzene bis-msb, benzene, p-bis o-methylstyryl, 1,4-di-2-methylstyryl benzene, 2-menthylstyrene, 1-methyl-2-2-4-2-2-methylphenyl ethenyl phenyl ethenyl benzene
PubChem CID 5378735
IUPAC Name 1,4-bis[(E)-2-(2-methylphenyl)ethenyl]benzene
SMILES CC1=CC=CC=C1C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3C

Specifications

Melting Point 179°C
Quantity 5 g
Formula Weight 310.44
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(2-methylstyryl)benzene [Solute for Liquid Scintillation Counting]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.