accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNH2H3COFFF

CAS RN 132861-87-1

IUPAC Name: 4,5-dimethyl-3-[3-(trifluoromethyl)benzoyl]thiophen-2-amine
Synonyms: 4,5-dimethyl-3-[3-(trifluoromethyl)benzoyl]thiophen-2-amine
Molecular Weight (g/mol): 299.31
Molecular Formula: C14H12F3NOS
InChi Key: KKDKAWKYGCUOGR-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C(=O)C2=CC=CC(=C2)C(F)(F)F)=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)