accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHOSOOCH3OHCH3

CAS RN 13288-70-5

IUPAC Name: 4-(4-hydroxy-3,5-dimethylbenzenesulfonyl)-2,6-dimethylphenol
Synonyms: 4-(4-hydroxy-3,5-dimethylbenzenesulfonyl)-2,6-dimethylphenol
Molecular Weight (g/mol): 306.38
Molecular Formula: C16H18O4S
InChi Key: SUCTVKDVODFXFX-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1O)S(=O)(=O)C1=CC(C)=C(O)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results