Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone 98.0+%, TCI America™
SDP

Catalog No. B1577500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1577500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1577500G Supplier TCI America Supplier No. B1577500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13288-70-5
Molecular Formula C16H18O4S
Molecular Weight (g/mol) 306.376
MDL Number MFCD00036032
InChI Key SUCTVKDVODFXFX-UHFFFAOYSA-N
Synonym Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone
PubChem CID 83310
IUPAC Name 4-(4-hydroxy-3,5-dimethylphenyl)sulfonyl-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)S(=O)(=O)C2=CC(=C(C(=C2)C)O)C

Specifications

Melting Point 297°C
Color White-Yellow
Quantity 500 g
Formula Weight 306.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-hydroxy-3,5-dimethylphenyl) Sulfone
EINECSNumber (3)-2169
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.