accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHOO(S)(S)NHOOHOO

CAS RN 13291-61-7

IUPAC Name: 2-{[(1S,2S)-2-[bis(carboxymethyl)amino]cyclohexyl](carboxymethyl)amino}acetic acid
Synonyms: {[(1S,2S)-2-[bis(carboxymethyl)amino]cyclohexyl](carboxymethyl)amino}acetic acid
Molecular Weight (g/mol): 346.34
Molecular Formula: C14H22N2O8
InChi Key: FCKYPQBAHLOOJQ-UWVGGRQHSA-N
SMILES: OC(=O)CN(CC(O)=O)[C@H]1CCCC[C@@H]1N(CC(O)=O)CC(O)=O
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results