Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid, Monohydrate, Macron Fine Chemicals™

Catalog No. MK2357100 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-2357-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-2357-100 Supplier Avantor Supplier No. 235757
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13291-61-7
Molecular Formula C14H22N2O8
Molecular Weight (g/mol) 346.336
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-N
Synonym 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid, trans-cyclohexane-1,2-dinitrilotetraacetic acid, acetic acid, 1,2-cyclohexylenedinitrilo tetra-, trans, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, trans, 2,2',2,2'-1r,2r-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid, rel-2,2',2,2'-1r,2r-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid, trans-1,2-cyclohexanediaminetetraacetic acid, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid, glycine, n,n'-1r,2r-1,2-cyclohexanediylbis n-carboxymethyl-, rel
PubChem CID 2723845
IUPAC Name 2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Quantity 100 g
Formula Weight 364.36
Percent Purity 98% min.
Grade ACS
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.