Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indigo Carmine, Harleco™, MilliporeSigma™

Catalog No. M24512 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24512 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24512 Supplier MilliporeSigma Supplier No. 24512

missing translation for 'validMainframeMsg'

Certified Biological Stain, Used for staining cytoplasm, negri bodies and connective tissue. can be mixed with Eosin Y as a counterstain to hematoxylin for staining vaginal smears

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13291-61-7
Molecular Formula C14H22N2O8
Molecular Weight (g/mol) 346.336
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-N
Synonym Carmine Blue
PubChem CID 2723845
IUPAC Name 2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Solubility Information 1g/L
Formula Weight 466.36
Packaging Glass Bottle
Color Blue
pH 7 (10 g/L, H2O, 20°C)
Quantity 25 g
Physical Form Lustrous Powder
EINECSNumber 212-728-8
RTECSNumber DU3000000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.