accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSOOHNNHSOOClClCl

CAS RN 133-67-5

IUPAC Name: 6-chloro-3-(dichloromethyl)-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
Synonyms: trichlormethiazide trichlormetazid trichlormas esmarin trichlormethiazide W/ reserpine trichlormethiazide preparation diurazida jones brand of trichloromethiazide chlopolidine tachionin
Molecular Weight (g/mol): 380.64
Molecular Formula: C8H8Cl3N3O4S2
InChi Key: LMJSLTNSBFUCMU-UHFFFAOYNA-N
SMILES: NS(=O)(=O)C1=CC2=C(NC(NS2(=O)=O)C(Cl)Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (5)
Grade 
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (1)
  • (9)
  • (3)
  • (5)
  • (4)
  • (1)
  • (4)
  • (5)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)